missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein L6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA5114867
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Antibody detects endogenous levels of total APOL6.
This gene is a member of the apolipoprotein L gene family. The encoded protein is found in the cytoplasm, where it may affect the movement of lipids or allow the binding of lipids to organelles.Spezifikation
Apolipoprotein L6 | |
Polyclonal | |
Unconjugated | |
APOL6 | |
APOL6; apolipoprotein L, 6; Apolipoprotein L6; apolipoprotein L-VI; APOLVI; APOL-VI; DKFZp667M075; FLJ38562; FLJ90164; MGC57495; UNQ3095/PRO21341 | |
Rabbit | |
Affinity chromatography | |
RUO | |
80830 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q9BWW8 | |
APOL6 | |
A synthesized peptide derived from human APOL6(Accession Q9BWW8), corresponding to amino acid residues M1-P47. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |