Learn More
Novus Biologicals™ Apolipoprotein L2 Overexpression Lysate
Marke:  Novus Biologicals™ NBL1-07624
Artikelnummer. 18268592
  • 820.00 CHF / 0.10 Milligramm
Verfügbar ab: 10-01-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
The Apolipoprotein L2 Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Spezifikation
Spezifikation
The lysate was created in HEK293T cells, using plasmid ID RC22585 and based on accession number NM_3882. The protein contains a C-terminal DDK tag.
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
Western Blot
RIPA buffer
Homo sapiens apolipoprotein L, 2 (APOL2), transcript variant alpha, mRNA.
Native
Sonderangebote
Nur für Forschungszwecke