Learn More
Novus Biologicals™ Apolipoprotein D Overexpression Lysate
Marke:  Novus Biologicals™ NBL1-07620
Artikelnummer. 18210213
  • 820.00 CHF / 0.10 Milligramm
Verfügbar ab: 10-01-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
The Apolipoprotein D Overexpression Lysate is derived from human. It has been validated for the following applications: Western Blot.
Spezifikation
Spezifikation
The lysate was created in HEK293T cells, using plasmid ID RC253 and based on accession number NM_1647. The protein contains a C-terminal DDK tag.
0.1 mg
Protein
-80°C. Avoid freeze-thaw cycles.
Native
Western Blot
RIPA buffer
Homo sapiens apolipoprotein D (APOD), mRNA.
ApoD, Apo-D, apolipoprotein D
Sonderangebote
Nur für Forschungszwecke