missing translation for 'onlineSavingsMsg'
Learn More

Apolipoprotein C2 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Thermo Scientific PA5116769

Artikelnummer. 17209273

  • 490.00 CHF / 100 Mikroliter
Verfügbar ab: 21-01-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Beschreibung

Beschreibung

Positive Samples: Rat liver, Rat plasma Immunogen sequence: MGTRLLPALF LVLLVLGFEV QGTQQPQQDE MPSPTFLTQV KESLSSYWES AKTAAQNLYE KTYLPAVDEK LRDLYSKSTA AMSTYTGIFT DQVLSVLKGE E

Apolipoproteins are protein components of plasma lipoproteins. The apolipoprotein C gene family encodes four homologous proteins designated apoC-I to -IV, which specifically modulate the metabolism of triglyceride-rich lipoproteins. The human apoC-I gene maps to chromosome 19q13.2 and is expressed primarily in the liver where it is activated when monocytes differentiate into macrophages. The human apoC-II gene maps to chromosome 19q13.2 and encodes a 79 amino acid single chain protein that is a necessary cofactor for the activation of lipoprotein lipase, the enzyme that hydrolyzes triglycerides in plasma and transfers the fatty acids to tissues.
TRUSTED_SUSTAINABILITY
Spezifikation

Spezifikation

Apolipoprotein C2
Polyclonal
Unconjugated
APOC2
APC 2; APC2; APO C2; Apo CII; APOC 2; ApoC II; Apoc2; APOC2 protein; ApoCII; APO-CII; apoC-II; Apolipoprotein C II; apolipoprotein C2; Apolipoprotein CII; Apolipoprotein C-II; ApolipoproteinCII; MGC75082; ProapoC-II; Proapolipoprotein C-II; RGD1560725
Rabbit
Affinity chromatography
RUO
292697, 344
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
1.65 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P02655
APOC2
Recombinant fusion protein containing a sequence corresponding to amino acids 1-101 of human APOC2 (NP_0004742)
100 μL
Primary
Human, Rat
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote