missing translation for 'onlineSavingsMsg'
Learn More

Apolipoprotein C1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

162.00 CHF - 315.00 CHF

Spezifikation

Antigen Apolipoprotein C1
Konzentration 0.5 mg/mL
Anwendungen Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Klassifikation Polyclonal
Konjugat Unconjugated
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
17909862
Unterlagen ansehen
Invitrogen
PA5145261
200 μL
315.00 CHF
200 Mikroliter
Verfügbar ab: 21-01-2025
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
17909472
Unterlagen ansehen
Invitrogen
PA5145262
20 μL
162.00 CHF
20 Mikroliter
Verfügbar ab: 21-01-2025
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

Apolipoproteins are protein components of plasma lipoproteins. The apolipoprotein C gene family encodes four homologous proteins designated apoC-I to -IV, which specifically modulate the metabolism of triglyceride-rich lipoproteins. The human apoC-I gene maps to chromosome 19q13.2 and is expressed primarily in the liver where it is activated when monocytes differentiate into macrophages. The human apoC-II gene maps to chromosome 19q13.2 and encodes a 79 amino acid single chain protein that is a necessary cofactor for the activation of lipoprotein lipase, the enzyme that hydrolyzes triglycerides in plasma and transfers the fatty acids to tissues. The human apoC-III gene maps to chromosome 11q23 and encodes a protein that may delay catabolism of triglyceride-rich particles by inhibiting lipoprotein lipase and hepatic lipase. The human apoC-IV gene maps to chromosome 19q13.2 and encodes a 127 amino acid protein that is primarily expressed in the liver.
Spezifikation

Spezifikation

Apolipoprotein C1
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Rabbit
Human, Mouse
P02654, P34928
11812, 341
Recombinant Human APOC1 protein, Ser31-Ser83 (Accession #P02654)
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
0.5 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.05% Proclin 300, pH 7.4
ALPCI, Apoc1, Apo-CI, ApoC-I, Apo-CIB, ApoC-IB, apolipoprotein C1, Apolipoprotein C-I, apolipoprotein C-I variant I, apolipoprotein C-I variant II, Liver regeneration-related protein LRRG04, LRRG04, Truncated apolipoprotein C-I
APOC1
IgG
Protein A
APOC1
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications