missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Apolipoprotein C1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Invitrogen PA5145499
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Apolipoproteins are protein components of plasma lipoproteins. The apolipoprotein C gene family encodes four homologous proteins designated apoC-I to -IV, which specifically modulate the metabolism of triglyceride-rich lipoproteins. The human apoC-I gene maps to chromosome 19q13.2 and is expressed primarily in the liver where it is activated when monocytes differentiate into macrophages. The human apoC-II gene maps to chromosome 19q13.2 and encodes a 79 amino acid single chain protein that is a necessary cofactor for the activation of lipoprotein lipase, the enzyme that hydrolyzes triglycerides in plasma and transfers the fatty acids to tissues. The human apoC-III gene maps to chromosome 11q23 and encodes a protein that may delay catabolism of triglyceride-rich particles by inhibiting lipoprotein lipase and hepatic lipase. The human apoC-IV gene maps to chromosome 19q13.2 and encodes a 127 amino acid protein that is primarily expressed in the liver.Spezifikation
Apolipoprotein C1 | |
Polyclonal | |
Unconjugated | |
APOC1 | |
ALPCI, Apoc1, Apo-CI, ApoC-I, Apo-CIB, ApoC-IB, apolipoprotein C1, Apolipoprotein C-I, apolipoprotein C-I variant I, apolipoprotein C-I variant II, Liver regeneration-related protein LRRG04, LRRG04, Truncated apolipoprotein C-I | |
Rabbit | |
Protein A | |
RUO | |
25292, 476437 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% Proclin 300, pH 7.4 | |
P19939, P56595 | |
APOC1 | |
Recombinant Dog APOC1 protein, Ile30-Ser88 (Accession #P56595) | |
200 μL | |
Primary | |
Canine, Rat | |
Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur