missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPP1, Human, Matched Antibody Pair (AP42), Abnova™
To detect and quantify protein level of human
Marke: Abnova H00006696-AP42.1Set
Includes: Capture antibody: mouse monoclonal anti-SPP1 (100μg) , Detection antibody: biotinylated mouse monoclonal anti-SPP1, IgG2a Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Spezifikation
SPP1 | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human SPP1. | |
BNSP/BSPI/ETA-1/MGC110940/OPN | |
Mouse | |
RUO | |
6696 | |
Human | |
Liquid |
ELISA | |
Biotin | |
SPP1 | |
SPP1 | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-SPP1 (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-SPP1, IgG2a Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
For Research Use Only