missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-MAPK3, ELK1 Protein Protein Interaction Antibody Pair, Abnova™
Marke: Abnova DI0280.1Set
Includes: MAPK3 rabbit purified polyclonal antibody (20μg); ELK1 mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Beschreibung
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the MAPK3 protein, and the other against the ELK1 protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Spezifikation
MAPK3 | |
Monoclonal/Polyclonal | |
MAPK3 | |
MAPK3 | |
1 Set | |
Primary | |
Antibody pair set content: 1. MAPK3 rabbit purified polyclonal antibody (20 ug) 2. ELK1 mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |
In situ PLA | |
Unconjugated | |
ERK1/HS44KDAP/HUMKER1A/MGC20180/P44ERK1/P44MAPK/PRKM3 | |
Mouse, Rabbit | |
RUO | |
5595/2002 | |
Human | |
MAPK3 & ELK1 Protein Protein Interaction Antibody Pair |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur