missing translation for 'onlineSavingsMsg'
Learn More
Learn More
anti-IL32, Human, IP-WB Antibody Pair, Abnova™
For immunoprecipitation and to detect the precipitated protein in western blot.
Marke: Abnova H00009235-PW1.1Set
Additional Details : Gewicht : 0.02000kg
Includes: Antibody pair for IP: rabbit polyclonal anti-IL32 (300μL), Antibody pair for WB: mouse polyclonal anti-IL32 (50μL)
Spezifikation
IL32 | |
Monoclonal/Polyclonal | |
This IP-WB antibody pair set comes with one antibody for immunoprecipitation and another to detect the precipitated protein in western blot. | |
IL-32alpha/IL-32beta/IL-32delta/IL-32gamma/NK4/TAIF/TAIFa/TAIFb/TAIFc/TAIFd | |
Mouse, Rabbit | |
RUO | |
9235 | |
Human | |
Liquid |
Immunoprecipitation, Western Blot | |
Unconjugated | |
IL32 | |
IL32 | |
1 Set | |
Primary | |
Antibody pair set content: 1. Antibody pair for IP: rabbit polyclonal anti-IL32 (300 ul) 2. Antibody pair for WB: mouse polyclonal anti-IL32 (50 ul) | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
For Research Use Only