missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IL1RN, Human, Matched Antibody Pair (AP42), Abnova™
To detect and quantify protein level of human
Marke: Abnova H00003557-AP42.1Set
Includes: Capture antibody: mouse monoclonal anti-IL1RN (100μg) , Detection antibody: biotinylated mouse monoclonal anti-IL1RN, IgG1 Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Spezifikation
IL1RN | |
Monoclonal/Polyclonal | |
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human IL1RN. | |
ICIL-1RA/IL-1ra3/IL1F3/IL1RA/IRAP/MGC10430 | |
Mouse | |
RUO | |
3557 | |
Human | |
Liquid |
ELISA | |
Biotin | |
IL1RN | |
IL1RN | |
1 Set | |
Primary | |
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-IL1RN (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-IL1RN, IgG1 Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur