Learn More
IL13, Human, Matched Antibody Pair (AP43), Abnova™
To detect and quantify protein level of human
Marke:  Abnova H00003596-AP43.1Set
Artikelnummer. 16132254
  • 872.00 CHF / 1 Set
Verfügbar ab: 07-01-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Zum Warenkorb hinzufügen
Includes: Capture antibody: mouse monoclonal anti-IL13 (100μg) , Detection antibody: biotinylated mouse monoclonal anti-IL13, IgG2a Kappa (50μg) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Spezifikation
IL13
Monoclonal/Polyclonal
This antibody pair set comes with matched antibody pair to detect and quantify protein level of human IL13.
ALRH/BHR1/IL-13/MGC116786/MGC116788/MGC116789/P600
Mouse
RUO
3596
Human
Liquid
ELISA
Biotin
IL13
IL13
1 Set
Primary
Antibody pair set content: 1. Capture antibody: mouse monoclonal anti-IL13 (100 ug) 2. Detection antibody: biotinylated mouse monoclonal anti-IL13, IgG2a Kappa (50 ug) *Reagents are sufficient for at least 3-5 x 96 well plates using recommended protocols.
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use.