Learn More
anti-FCGR3A, FCGR1A Protein Protein Interaction Antibody Pair, Abnova™
Marke: Abnova DI0195.1Set
Additional Details : Gewicht : 0.00010kg
Includes: FCGR3A rabbit purified polyclonal antibody (20μg); FCGR1A mouse monoclonal antibody (40μg); Reagents are sufficient for at least 30-50 assays using recommended protocols
Beschreibung
This protein protein interaction antibody pair set comes with two antibodies to detect the protein-protein interaction, one against the FCGR3A protein, and the other against the FCGR1A protein for use in in situ Proximity Ligation Assay.
Host Species: Rabbit, Mouse
Species Reactivity: Hu
In situ PLA
Spezifikation
FCGR3A | |
Monoclonal/Polyclonal | |
FCGR3A | |
FCGR3A | |
1 Set | |
Primary | |
Antibody pair set content: 1. FCGR3A rabbit purified polyclonal antibody (20 ug) 2. FCGR1A mouse monoclonal antibody (40 ug) *Reagents are sufficient for at least 30-50 assays using recommended protocols. | |
Store reagents of the antibody pair set at -20°C or lower. Please aliquot to avoid repeated freeze thaw cycle. Reagents should be returned to -20°C storage immediately after use. | |
Purified |
In situ PLA | |
Unconjugated | |
CD16/CD16A/FCG3/FCGR3/FCGRIII/FCR-10/FCRIII/FCRIIIA/IGFR3 | |
Mouse, Rabbit | |
RUO | |
2214/2209 | |
Human | |
FCGR3A & FCGR1A Protein Protein Interaction Antibody Pair |