Learn More
anti-Cyclin D1/Bcl-1, Clone: SP4, Epredia™ LabVision
The Epredia™ Cyclin D1/Bcl-1, Rabbit Monoclonal Antibody is part of a comprehensive antibody portfolio. Studies show rabbit monoclonal antibodies are more sensitive and specific than other available markers.
Marke: Epredia RM-9104-R7
579.92 CHF gültig bis 2024-12-31
Benutzen Sie den Promotions-Code "21628" um Ihren Promotions-Preis zu erhalten.
Beschreibung
Cyclin D1 or PRAD-1 or bcl-1 is one of the key cell cycle regulators, and functions in association with cdk4 and/or cdk6 by phosphorylating the Rb protein. It is a putative proto-oncogene overexpressed in a wide variety of human neoplasms including mantle cell lymphomas (MCL).
Host Species: Rabbit
Clone: SP4
Isotype: IgG
Species Reactivity: Human, Mouse, Rat. Others not tested.
Epitope: C-terminus
Immunogen: A synthetic peptide from C-terminus of human cyclin D1
Molecular Weight: 36kDa
Positive Control: MAD109 cells, breast CA or mantle cell lymphoma
Cellular Localization: Nuclear
Clone SP4 is excellent for staining of formal/paraffin tissue.
†USA: IVD; Int'l: IVD
Immunohistochemistry (Paraffin), Western Blotting
Spezifikation
Cyclin D1/Bcl-1 | |
SP4 | |
Unconjugated | |
A synthetic peptide from C-terminus of human cyclin D1 | |
IVD | |
Primary | |
Prediluted |
Monoclonal | |
n/a | |
Rabbit | |
7 mL | |
Cell Cycle | |
Human, Mouse, Rat | |
IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.