Learn More
CD62P Mouse anti-Human, Unlabeled, Clone: AK-4, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 555522
Additional Details : Gewicht : 0.01000kg
Beschreibung
The AK-4 monoclonal antibody specifically binds to CD62P. CD62P is a 140 kDa type I transmembrane glycoprotein that is also known as P-Selectin, Platelet activation-dependent granule membrane protein (PADGEM), or GMP-140. P-Selectin is stored in the α-granules of platelets and the Weibel-Palade bodies of endothelial cells, and is rapidly transported to the plasma membrane upon activation. P-Selectin is thought to mediate the initial adhesive interactions of neutrophils and monocytes with endothelium in inflammatory responses, and of activated platelets to neutrophils and monocytes in hemostasis.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Spezifikation
CD62P | |
Monoclonal | |
0.5mg/mL | |
P-Selectin; GMP-140; PADGEM | |
Mouse | |
0.1 mg | |
Immunology | |
Human | |
IgG1 κ |
Flow Cytometry | |
AK-4 | |
Unconjugated | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
For Research Use Only.