Learn More
BrdU with DNase Mouse anti-Human, FITC, Clone: B44, BD
Mouse Monoclonal Antibody
Marke: BD Biosciences 340649
Beschreibung
The Anti-BrdU antibody, clone B44, is derived from hybridization of Sp2/0-Ag14 mouse myeloma cells with spleen cells from BALB/c mice immunized with iodouridine-conjugated ovalbumin.
Bromodeoxyuridine (BrdU) is a uridine derivative that can be incorporated specifically into DNA in place of thymidine. Anti-BrdU identifies BrdU (but not thymidine) in single-stranded DNA, free BrdU, or BrdU coupled to a protein carrier. The antibody also reacts with iodouridine. DNase-I cleaves each strand of DNA at random and permits the binding of Anti-BrdU antibody to its antigen (BrdU).
Spezifikation
BrdU with DNase | |
Monoclonal | |
10μg/mL | |
Balanced salt solution with DNase, gelatin, and 0.1% sodium azide. | |
Iodouridine | |
50 Tests | |
Primary | |
Store vials at 2-8°C. Conjugated forms should not be frozen and should be protected from prolonged exposure to light. Each reagent is stable for the period shown on the bottle label when stored as directed. |
Flow Cytometry, Immunocytochemistry | |
B44 | |
FITC | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
For Research Use Only.