Learn More
Ansell MICRO-TOUCH™ Nitrile Sterile Gloves
Artikelnummer. 17843772
Click to view available options
Größe:
S
M
L
XL
Nitrile, sterile pair packed, powder free examination glove featuring extended cuff.
  • Sterile glove made from a synthetic, nitrile formulation
  • High elasticity with barrier protection and puncture resistance
  • Ideal solution for a range of sterile examination (non-surgical) procedures
  • Packed as a pair of gloves per pack
  • 12 in. extended cuff for additional protection
  • Textured fingers provide extra grip and reliability
  • Gamma sterilized minimum 25kGγ to provide sterility
  • Tested for use with chemotherapy drugs
Spezifikation
Farbe Blau
Einwegprodukt Zur einmaligen Verwendung
Material Nitril
Größe XL
Modell 6034154
Länge (angloamerikanisch) 11,6 in.
Länge (metrisch) 29.4 cm