Learn More
Amphiregulin Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech CL488-16036-100UL
Additional Details : Gewicht : 0.00020kg
Beschreibung
The Amphiregulin antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Amphiregulin. This antibody recognizes human antigen. The Amphiregulin antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Spezifikation
Amphiregulin | |
Polyclonal | |
Rabbit | |
Primary | |
Human |
Immunofluorescence | |
CoraLite 488 | |
100 μL | |
374 | |
IgG |