missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminiumoxid, EcoChrom™, MP Biomedicals™
Click to view available options
Menge:
50 kg
Spezifikation
Spezifikation
Chemischer Name oder Material | Aluminiumoxid |
CAS | 1344-28-1 |
Menge | 50 kg |
Summenformel | Al2O3 |
MDL-Nummer | MFCD00003424 |
Synonym | Aluminiumoxid, Alpha-Aluminiumoxid, Aloxit, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum |
InChI-Schlüssel | PNEYBMLMFCGWSK-UHFFFAOYSA-N |
SMILES | [O--].[O--].[O--].[Al+3].[Al+3] |
IUPAC-Name | Dialuminium;sauerstoff(2-) |
Molekulargewicht (g/mol) | 101.96 |
Mehr anzeigen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'