Learn More
Wesbart™ Aluminum Horizontal Freezer Rack
Wesbart™ Aluminum Horizontal Freezer Racks are front loading with trays for quick access to cryoboxes and other stored items.
Marke:  Wesbart™ HC2706
 Lernen Sie weitere Varianten dieses
Artikelnummer. 15884732
  • 166.00 CHF / 1 Stück
Verfügbar ab: 14-02-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
  • Aluminum front access storage rack for tray units (ordered separately, Cat. No. 17311849) for upright freezers
  • For standard cryobox or other freezer storage
  • Only one tray has to be removed from the rack at a time for sample access, minimizing temperature impact on remaining items
  • Range of sizes and capacities available
Spezifikation
Spezifikation
Freezer Storage Rack
Aluminum
Tiefkühlschränke
Loose Items or Cryoboxes
1 Tray/Rack
560 x 140 x 290 mm