missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminiumoxid G-TLC, EcoChrom™, MP Biomedicals™
Click to view available options
Menge:
1 kg
Spezifikation
Spezifikation
Chemischer Name oder Material | Aluminiumoxid G-TLC |
CAS | 1344-28-1 |
Farbe | Rosa bis Weiß |
Physikalische Form | Pulver |
Menge | 1 kg |
Summenformel | Al2O3 |
MDL-Nummer | MFCD00003424 |
Synonym | Aluminiumoxid, Alpha-Aluminiumoxid, Aloxit, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum |
InChI-Schlüssel | PNEYBMLMFCGWSK-UHFFFAOYSA-N |
SMILES | [O--].[O--].[O--].[Al+3].[Al+3] |
Mehr anzeigen |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'