missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alumina, Activity: II-III, MP EcoChrome™
Click to view available options
MP EcoChrome™ Alumina, Activity: II-III, 500 g
Beschreibung
MP EcoChrom™ Alumina, Activity II-III is an economical adsorbents of medium activity.
Is often used where:
- Activated charcol can not be used due to its organic nature
- The cation exchange properties of basic alumina are desired
MP EcoChrom™ Alumina, Activity II-III is:
- Suitable for the development of separation methods and for technical separations where a custom-made adsorbents for special separation is not available
- An oxide of aluminum, occurring in nature as various minerals such as: bauxite, corundum, etc.
- Used as:
- An adsorbent
- A desiccating agent and a catalyst, in the manufacture of dental cements and refractories.
Application Notes:
MP EcoChrom™ Alumina catalyses a variety of reactions that are useful industrially.
It is used:
- To remove water from gas streams
- For its hardness and strength
- As an abrasive, including as a much less expensive substitute for industrial diamond. Many types of sandpaper use aluminium oxide crystals.
- In chemistry laboratories, alumina is a medium for chromatography, available in basic (pH 9.5), acidic (pH 4.5 when in water) and neutral formulations, as well as for alcohols, hydrocarbons, steroids, alkaloids and natural pigments.
- Neutral alumina, (less active than the basic form) is useful for separation of:
- Aldehydes
- Ketones
- Quinones
- Esters
- Lactones
- Glucosides
- Acidic alumina, is the weakest and least often used type and is useful for separation of acid pigments and strong acids. It can cause:
- Polymerization
- Condensation
- Dehydration reactions
- Neither acetone nor ethyl acetate should be used as eluants, the latter being subject to saponification
- A dosimeter for radiation protection and therapy applications for its optically stimulated luminescence properties
- Neutral alumina, (less active than the basic form) is useful for separation of:
Usage Statement: Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Key Applications:
- Chromatography
- Adsorbants
- TLC
Spezifikation
Spezifikation
Siedepunkt | ca. 2980°C (Lit.) |
Chemischer Name oder Material | Aluminum oxide |
Schmelzpunkt | ca. 2072°C (Lit.) |
CAS | 1344-28-1 |
pH | 10 |
Physikalische Form | Powder |
Menge | 500 g |
Summenformel | Al2O3 |
MDL-Nummer | MFCD00003424 |
InChI-Schlüssel | PNEYBMLMFCGWSK-UHFFFAOYSA-N |
Mehr anzeigen |
Sofern nicht anders angegeben, sind die Produkte von MP Biomedical nur zur Forschung oder Weiterverarbeitung, aber nicht zur direkten Anwendung am Menschen bestimmt.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'