missing translation for 'onlineSavingsMsg'
Learn More

Aluminiumoxid A, Aktivität: I, EcoChrom™, MP Biomedicals™

Artikelnummer. 17807034
Click to view available options
Menge:
500 g
5 kg
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

MP EcoChrom™ Alumina A catalyses a variety of reactions that are useful industrially.

  • Alumina A is widely used to remove water from gas streams.
  • Also used for its hardness and strength.
  • It is widely used as an abrasive, including as a much less expensive substitute for industrial diamond.
  • Many types of sandpaper use aluminium oxide crystals.
  • In chemistry laboratories, alumina is a medium for chromatography, available in basic (pH 9.5), acidic (pH 4.5 when in water) and neutral formulations, as well as for alcohols, hydrocarbons, steroids, alkaloids and natural pigments.
  • Neutral alumina, (less active than the basic form) is useful for separation of aldehydes, ketones, quinones, esters, lactones, and glucosides.
  • Acidic alumina, is the weakest and least often used type.
  • Alumina is useful for separation of acid pigments and strong acids.
  • It can cause polymerization, condensation and dehydration reactions.
  • Neither acetone nor ethyl acetate should be used as eluants, the latter being subject to saponification.
  • It is used as a dosimeter for radiation protection and therapy applications for its optically stimulated luminescence properties.
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 1344-28-1
Summenformel Al2O3
Molekulargewicht (g/mol) 101.96
MDL-Nummer MFCD00003424
InChI-Schlüssel PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC-Name Dialuminium(3+) Trioxidandiid
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Spezifikation

Chemischer Name oder Material Alumina A
Namenshinweis Activity: I, Activity acc. To Brockmann: 23 l/mm
Farbe Rosa bis Weiß
Physikalische Form Free-flowing Powder
Menge 500 g
Formelmasse 101.96
Dichte 3.94 g/cm3 (at 20°C)

Sofern nicht anders angegeben, sind die Produkte von MP Biomedical nur zur Forschung oder Weiterverarbeitung, aber nicht zur direkten Anwendung am Menschen bestimmt. Weitere Informationen erhalten Sie von unserem Kundendienst.

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'