missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Agarose UHR
For DNA/RNA fragments 5-500bp.
656.00 CHF - 2760.00 CHF
Spezifikation
Chemischer Name oder Material | Agarose UHR |
---|---|
CAS | 9012-36-6 |
Güte | Molecular Biology |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
30017991
|
Melford
A20135-500.0 |
500 g |
2760.00 CHF
500 Gramm |
Verfügbar ab: 14-01-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
30017990
|
Melford
A20135-100.0 |
100 g |
656.00 CHF
100 Gramm |
Verfügbar ab: 14-01-2025
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
For DNA/RNA fragments 5-500bp.
For DNA/RNA fragments 5-500bp. Ideal for primer size. DNAse and RNase free at 3% concentration, Agarose Ultra High Resolution (UHR) presents a resolution of DNA fragments similar to gels made with polyacrylamide at concentrations of 8%. Gels with Agarose UHR are easier to prepare and are absolutely free of neurotoxic dangers inherent to polyacrylamide. While Agarose UHR can be dissolved by careful adherence to microwaving procedures, Agarose UHR gels are best prepared by autoclaving.Spezifikation
Agarose UHR | |
9012-36-6 | |
Molecular Biology |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title