missing translation for 'onlineSavingsMsg'
Learn More

Melford Agarose SS

For DNA/RNA fragments >1000 bp.

Brand:  Melford A20097-500.0

 View more versions of this product

Product Code. 30017988

  • 810.00 CHF / 500 Gramm
Verfügbar ab: 14-01-2025
Sie sich ein, um den Lagerbestand zu sehen
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

For DNA/RNA fragments >1000 bp.

For DNA/RNA fragments >1000 bp. Ideal for separating larger molecules. DNAse/RNase free.

Gel Strength, 1%: >1800 g/cm2

Gel Strength, 1.5%: >3200 g/cm2

Gelling Temperature: 35-37°C

Melting Temperature: 87-89°C

Sulfate: <0.12%

EEO: <0.13%

Specifications

Specifications

Agarose SS
9012-36-6
500 g
Molecular Biology
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.