Learn More
AffiniPure Goat anti-Rabbit, Biotin, Polyclonal, Proteintech
Goat Polyclonal Antibody
Marke: Proteintech SA00004-2-200UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
AffiniPure Polyclonal antibody specifically detects AffiniPure in Human, Mouse, Rat samples. It is validated for ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry.
Spezifikation
AffiniPure | |
Polyclonal | |
Biotin | |
200 μL | |
Human, Mouse, Rat | |
IgG |
ELISA, Western Blot, Immunohistochemistry, Immunocytochemistry | |
0.3 mg/mL | |
Goat | |
Secondary | |