Learn More
BioTek™ 96-tube Rack for Test Tubes
96-tube Rack for 12-13 mm Test Tubes is for use with BioTek™ Microplate Washers and Readers
Marke: BioTek™ 7110537
Additional Details : Gewicht : 0.50000kg
Spezifikation
Tube Rack | |
96 Test Tubes | |
BioTek™ Microplate Washer and Reader Instruments | |
12 to 13 mm |