Learn More
SHOWA™ 477 Series Nitrile Coated Cold Resistant Gloves
Click to view available options
Größe:
M
L
XL
2XL
Beschreibung
Benifits
- Seamless knit
- Cold Resistant
- Water-resistant
- Flexible
- Abrasion-resistant
- Extra Grip
- Thermal insulation
- Form-fitting
- Impermeable
Features
- Foam grip
- Elasticated wrist cuff
- Ergonomic
- Foam nitrile palm dip
- Insulated
- Fully-coated nitrile
Trades & Applications
- Concreting
- Guttering and drain pipe
- Insulation
- Washing and cleaning
- Planting & repotting
- Bottling
- Water/Sewage treatment
- Plumbing
- Drainage, piping
- Shipping/Receiving
Spezifikation
Spezifikation
Zertifizierung/Konformität | CE Kat II, EN 388:2016, EN 511:2006 |
Bauweise | Nahtloser Strick |
Coating Material | Nitril |
Farbe | Blau, Schwarz |
Material | Nylon, Polyester |
Größe | M |
Auskleidungsmaterial | Acryl |
Beständig gegen | Kältebeständig, Wasserfest, Abriebfest |
Serie | 477 |