Learn More
TubeWriter 400 - Custom Rack Side Print for Any Labware
Custom racks made to exact specifications to print directly on side of unique labware
Marke:  TubeWriter PS-0000XX-EL-400-Custom
Artikelnummer. 30067431
  • 2965.00 CHF / 1 Stück
Verfügbar ab: 25-02-2025
Sie sich ein, um den Lagerbestand zu sehen
Entdecken Sie weitere Sonderangebote
Beschreibung
  • Custom racks desinged for the 400
  • Racks made to your exact specifications to directly print and label on the side of any labware
  • Rack will ensure correct alignment and holds labware securely
  • No time-consuming sticky labels or handwriting required
  • Racks designed for rapid switching between labware types
  • Print survives long-term storage at -80°C and liquid nitrogen
  • Print withstands the harshest lab conditions
  • Can print 2D or 3D barcodes, images and 5 lines of text
Spezifikation
Spezifikation
Labeling Rack