missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Nitro-N-(2-phenylethyl)benzamid, 97 %, Thermo Scientific™
203.00 CHF - 618.00 CHF
Chemische Identifikatoren
CAS | 62497-65-8 |
---|---|
Summenformel | C15H14N2O3 |
Molekulargewicht (g/mol) | 270.29 |
MDL-Nummer | MFCD00420506 |
InChI-Schlüssel | NPTZNBFQWMQHKP-UHFFFAOYSA-N |
Synonym | 4-nitro-n-2-phenylethyl benzamide, 4-nitro-n-phenethylbenzamide, 4-nitrophenyl-n-2-phenylethyl carboxamide, cambridge id 5176014, n-phenethyl-4-nitrobenzamide, 4-nitro-n-phenethyl-benzamide, benzamide, 4-nitro-n-2-phenylethyl |
PubChem CID | 230117 |
IUPAC-Name | 4-nitro-N-(2-phenylethyl)benzamide |
SMILES | [O-][N+](=O)C1=CC=C(C=C1)C(=O)NCCC1=CC=CC=C1 |
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemische Identifikatoren
62497-65-8 | |
270.29 | |
NPTZNBFQWMQHKP-UHFFFAOYSA-N | |
230117 | |
[O-][N+](=O)C1=CC=C(C=C1)C(=O)NCCC1=CC=CC=C1 |
C15H14N2O3 | |
MFCD00420506 | |
4-nitro-n-2-phenylethyl benzamide, 4-nitro-n-phenethylbenzamide, 4-nitrophenyl-n-2-phenylethyl carboxamide, cambridge id 5176014, n-phenethyl-4-nitrobenzamide, 4-nitro-n-phenethyl-benzamide, benzamide, 4-nitro-n-2-phenylethyl | |
4-nitro-N-(2-phenylethyl)benzamide |
Spezifikation
4-Nitro-N-(2-phenylethyl)benzamide | |
C15H14N2O3 | |
4-nitro-n-2-phenylethyl benzamide, 4-nitro-n-phenethylbenzamide, 4-nitrophenyl-n-2-phenylethyl carboxamide, cambridge id 5176014, n-phenethyl-4-nitrobenzamide, 4-nitro-n-phenethyl-benzamide, benzamide, 4-nitro-n-2-phenylethyl | |
[O-][N+](=O)C1=CC=C(C=C1)C(=O)NCCC1=CC=CC=C1 | |
270.29 | |
270.28 |
62497-65-8 | |
MFCD00420506 | |
NPTZNBFQWMQHKP-UHFFFAOYSA-N | |
4-nitro-N-(2-phenylethyl)benzamide | |
230117 | |
97% |
Sicherheit und Handhabung
GHS H-Satz
H315-H319-H335
Verursacht Hautreizungen.
Verursacht schwere Augenreizung.
Kann die Atemwege reizen.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
Gehäuse H315-H319-H335
missing translation for 'tsca' : Nein
Recommended Storage : Umgebungstemperaturen