missing translation for 'onlineSavingsMsg'
Learn More

3-Octanol, 97 %, Thermo Scientific Chemicals

Artikelnummer. 11461818
missing translation for 'orderingAttributeHoverText'
Menge:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Dieses Markenprodukt von Thermo Scientific Chemicals war ursprünglich Teil des Produktportfolios von Alfa Aesar. Einige Dokumente und Etiketteninformationen enthalten möglicherweise Angaben zur bisherigen Marke. Die ursprüngliche Produkt- oder Artikelnummer bei Alfa Aesar oder die SKU-Referenz wurde im Rahmen des Übergangs der Marke zu Thermo Scientific Chemicals nicht geändert.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 589-98-0
Summenformel C8H18O
Molekulargewicht (g/mol) 130.23
MDL-Nummer MFCD00004590
InChI-Schlüssel NMRPBPVERJPACX-UHFFFAOYNA-N
Synonym 3-octanol, octanol-3, amyl ethyl carbinol, ethyl-n-amylcarbinol, amylethylcarbinol, ethylamylcarbinol, 1-ethylhexanol, d-n-octanol, ethyl amyl carbinol, s-3-octanol
PubChem CID 11527
ChEBI CHEBI:80945
IUPAC-Name Octan-3-ol
SMILES CCCCCC(O)CC

Specifications

Chemischer Name oder Material 3-Octanol
Schmelzpunkt -45°C
Dichte 0.82
Siedepunkt 175°C to 176°C
Flammpunkt 68°C (154°F)
Geruch Pleasant
Brechungsindex 1.427
Menge 250 g
Beilstein 1719310
Formelmasse 130.23
Reinheit (%) 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.