CAS RN 101-55-3
CAS RN 101-55-3
IUPAC Name:
1-bromo-4-phenoxybenzene
Synonyme:
4-bromophenyl phenyl ether
Molekulargewicht (g/mol):
249.11
Summenformel:
C12H9BrO
InChi Key:
JDUYPUMQALQRCN-UHFFFAOYSA-N
SMILES:
BrC1=CC=C(OC2=CC=CC=C2)C=C1
1
–
1
von
1
Ergebnisse
1
4-Bromdiphenylether, 99 %, Thermo Scientific Chemicals
CAS: 101-55-3 Summenformel: C12H9BrO Molekulargewicht (g/mol): 249.107 MDL-Nummer: MFCD00000094 InChI-Schlüssel: JDUYPUMQALQRCN-UHFFFAOYSA-N Synonym: 4-bromodiphenyl ether,4-bromophenyl phenyl ether,4-bromophenoxybenzene,p-bromodiphenyl ether,p-phenoxybromobenzene,benzene, 1-bromo-4-phenoxy,p-bromophenyl phenyl ether,4-bromodiphenylether,p-bromophenoxybenzene,ether, p-bromophenyl phenyl PubChem CID: 7565 ChEBI: CHEBI:77421 IUPAC-Name: 1-brom-4-phenoxybenzol SMILES: C1=CC=C(C=C1)OC2=CC=C(C=C2)Br